Fluorescamine
500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 38183-12-9 Molecular Formula: C17H10O4 Formula Weight: 278.27 Physical Form: Powder Appearance: White to cream Odour: Odourless Melting Point: 155-158° MDL No.: MFCD00005928
Applications: For fluorescent detection of amino acids, peptides and proteins
Synonyms: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
- CAS Number:38183-12-9
- Formula:C₁₇H₁₀O₄
- MDL Number:MFCD00005928
- Molecular Weight:278.26 g/mol
- Merck Index:14,4158
- Melting Pt:155…158 °C
- Storage Temperature:Ambient
- EINECS:253-814-5
Documents
Specifications
Specifications
- Pk:500 mg



