Volumetric flasks, class B, blue markings
25 ml, 10/19, 110 mm, 0,08 ± ml
Catálogo # 612-3332
Fornecedor: SciLabware
Ir para:
Product Details & Documents
MBL®, borosilicate glass, with PE stopper.
- Marks and inscriptions in high contrast blue enamel
- Excellent temperature resistance
: Complies with ISO 1042 and DIN 12664
Especificações
- NS:10/19
- Capacity:25 ml
- Height:110 mm
- Seq:04
- Tolerance:0,08 ± ml



