Chloroparaffin C10-C13, 55,5% Cl 100 ng/µl 1 * 10 mL
EHERX23105500CY
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffers, ready to use
: WTW
Buffer solutions in sachets
: Mettler - Toledo