Order Entry
Portugal
Orders LinkContactUsLinkComponent
Comparators, Lovibond® Comparator System 2000+

Comparators, Lovibond® Comparator System 2000+

Nessleriser 2250 with stand
Catalog # 705-0344
Supplier:   Lovibond
Comparators, Lovibond® Comparator System 2000+
Catalog # 705-0344
Supplier:  Lovibond
Comparators, Lovibond® Comparator System 2000+
Comparators, Lovibond® Comparator System 2000+

Nessleriser 2250 with stand

Price
579,00 €
Log in for availability
1 * 1 unid.
Product Details & Documents

The Comparator System 2000+ is a precise and general purpose colorimetric system for water testing. It is a high quality instrument for visually matching the intensity of the colour of the sample with Lovibond glass filters. It includes an adjustable cell compartment for samples in cells of up to 40 mm in path length. Nessleriser 2150 and the 2250 are longer path length instruments for matching a column of a sample in a Nessler cylinder with Lovibond glass filters. They are designed for measuring concentrations that are below the sensitivity of the Comparator 2000+.

  • The integrated prism presents the glass standard discs and the coloured sample in the same field of vision, which allows for discrimination and matching of colours
  • The prism is ultrasonically sealed to prevent contamination of the optics
  • Each test disc is equipped with glass filters which guarantee a stability of colour, which do not fade and which can be changed easily without having to open the apparatus
  • Allows for the compensation of water samples that are turbid or tinted
  • Each comparator is available as a natural daylight option or can be used with the standardised Daylight 2000 lighting unit or a portable lighting unit
Specifications
  • Description:
    Nessleriser 2250 with stand
  • Seq:
    04
Accessories

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • LOVI230120
    QAC (Quaternary Ammonia Compounds)
    3/118
    2 - 20 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • LOVI230250
    Silica
    3/140
    0,1 - 1,0 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • LOVI231200
    Phosphate
    3/12
    0 - 80 mg/l
    Log in for availability
    1 * 1 unid. - 288,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • LOVI235330
    Bromine
    3/53C
    0,5 - 6,0 mg/l
    Log in for availability
    1 * 1 unid. - 288,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • LOVI281030
    pH
    NLC
    6,0 - 7,6 pH
    Log in for availability
    1 * 1 unid.

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • TINT243030
    Colour (Gardner/APHA)
    4/30AS
    1,0 - 9,0 pH
    Log in for availability
    1 * 1 unid. - 453,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • TINT243040
    Colour (Gardner/APHA)
    4/30BS
    10,0 - 18,0 pH
    Log in for availability
    1 * 1 unid. - 453,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • TINT246410
    AF332 EP colour scale
    4/64A
    R1 - R7
    Log in for availability
    1 * 1 unid. - 388,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • TINT246440
    AF332 EP colour scale
    4/64D
    BY1 - BY7
    Log in for availability
    1 * 1 unid. - 388,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • TINT246430
    AF332 EP colour scale
    4/64C
    B1 - B9
    Log in for availability
    1 * 1 unid. - 388,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • TINT246450
    AF332 EP colour scale
    4/64E
    GY1 - GY7
    Log in for availability
    1 * 1 unid. - 388,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0618
    pH
    2/1L
    8,0 - 9,6 pH
    Log in for availability
    1 * 1 unid. - 228,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0619
    Zinc
    3/102
    0 - 4,0 mg/l
    Log in for availability
    1 * 1 unid. - 228,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0620
    Nitrite
    3/103
    0,05 - 0,5 mg/l
    Log in for availability
    1 * 1 unid. - 288,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0165
    Copper
    3/110
    0 - 4,0 mg/l
    Log in for availability
    1 * 1 unid. - 288,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0167
    Ammonia
    3/112
    0 - 0,4 NH₄ mg/l
    Log in for availability
    1 * 1 unid. - 228,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0166
    Copper
    3/106
    0 - 1,0 mg/l
    Log in for availability
    1 * 1 unid. - 228,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0168
    Ammonia
    3/113
    0 - 1,0 N mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0621
    Iron
    3/116
    0,1 - 1,0 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0622
    Iron
    3/117
    1,0 - 10,0 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0623
    Nitrate
    3/124
    0,1 - 1,0 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0169
    Ammonia
    3/125
    0 - 10 N mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0170
    Hydrazine
    3/126
    0 - 0,5 mg/l
    Log in for availability
    1 * 1 unid. - 228,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 633-0171
    Aluminium
    3/127A
    0 - 0,5 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0625
    Phosphate
    3/133
    0 - 4,0 mg/l
    Log in for availability
    1 * 1 unid. - 310,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0628
    Molybdate
    3/138
    10 - 150 mg/l
    Log in for availability
    1 * 1 unid. - 244,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0629
    Silica
    3/139
    0,4 - 4,0 mg/l
    Log in for availability
    1 * 1 unid. - 288,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-0631
    Silica
    3/147
    1 - 10 mg/l
    Log in for availability
    1 * 1 unid. - 228,00 €

Product Information

  • Catalog #
    Test method
    Code
    Range
    Availability
    Price
  • 705-1039
    Molybdate
    3/162
    0 - 10 mg/l
    Log in for availability
    1 * 1 unid. - 230,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Special Nessler tube (determination of oxygen with disc NOE), AF 315
    Special Nessler tube (determination of oxygen with disc NOE), AF 315
    LOVI353150
    Log in for availability
    1 * 2 unid. - 235,00 €

Product Information

  • Catalog #
    Availability
    Price
  • DB424/S Glass cells square 13.5 mm path length with lid
    DB424/S Glass cells square 13.5 mm path length with lid
    634-8590
    Log in for availability
    1 * 5 unid. - 131,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Glass stirring rod, 12 cm
    Glass stirring rod, 12 cm
    LOVI364110
    Log in for availability
    1 * 1 unid. - 9,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 100 ml
    Dilution/shaker tube with lid, plastic, graduated, 100 ml
    LOVI385130
    Log in for availability
    1 * 1 unid. - 10,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Dilution/shaker tube with lid, plastic, graduated, 30 ml
    Dilution/shaker tube with lid, plastic, graduated, 30 ml
    LOVI385132
    Log in for availability
    1 * 1 unid. - 10,80 €

Product Information

  • Catalog #
    Availability
    Price
  • Cuvette stand for 10 round cuvettes, 16 mm Ø
    Cuvette stand for 10 round cuvettes, 16 mm Ø
    LOVI418957
    Log in for availability
    1 * 1 unid. - 45,20 €

Product Information

  • Catalog #
    Availability
    Price
  • W680/2.5 mm path length, fused optical glass rectangular cell
    W680/2.5 mm path length, fused optical glass rectangular cell
    705-0404
    Log in for availability
    1 * 1 unid. - 112,00 €

Product Information

  • Catalog #
    Availability
    Price
  • W680/5 mm path length, fused optical glass rectangular cell
    W680/5 mm path length, fused optical glass rectangular cell
    705-0401
    Log in for availability
    1 * 1 unid. - 112,00 €

Product Information

  • Catalog #
    Availability
    Price
  • W680/25 mm path length, fused optical glass rectangular cell
    W680/25 mm path length, fused optical glass rectangular cell
    705-0403
    Log in for availability
    1 * 1 unid. - 116,00 €

Product Information

  • Catalog #
    Availability
    Price
  • W680/40 mm path length, fused optical glass rectangular cell
    W680/40 mm path length, fused optical glass rectangular cell
    705-0566
    Log in for availability
    1 * 1 unid. - 116,00 €

Product Information

  • Catalog #
    Availability
    Price
  • AF306 Pair of Nessler tubes, 50 ml/113 mm
    AF306 Pair of Nessler tubes, 50 ml/113 mm
    TINT353060
    Log in for availability
    1 * 2 unid. - 32,10 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.