Aprotinin
Glass bottle for solids, 100 mg
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Cat. No.:A2132.0100
- Molecular formula:C₂₈₄H₄₃₂N₈₄O₇₉S₇
- Protein/peptide name:Aprotinin
- Storage conditions:Refrigerator
- Protein synonyms:Trypsin inhibitor, pancreatic basic
- MDL No.:MFCD00130541
- CAS No.:9087-70-1



