Order Entry
Portugal
Orders LinkContactUsLinkComponent
Aprotinin

Aprotinin

Glass bottle for solids, 25 mg
Catalog # A2132.0025
Supplier:   PanReac AppliChem
Aprotinin
Catalog # A2132.0025
Supplier:  PanReac AppliChem
CAS Number:  
Aprotinin

Glass bottle for solids, 25 mg

CAS Number:  
Price
108,00 €
Log in for availability
1 * 25 mg
Product Details & Documents

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Cat. No.:
    A2132.0025
  • Molecular formula:
    C₂₈₄H₄₃₂N₈₄O₇₉S₇
  • Protein/peptide name:
    Aprotinin
  • Storage conditions:
    Refrigerator
  • Protein synonyms:
    Trypsin inhibitor, pancreatic basic
  • MDL No.:
    MFCD00130541
  • CAS No.:
    9087-70-1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.