Order Entry
Portugal
Orders LinkContactUsLinkComponent
Forceps

Forceps

Tweezers, in PMP, Straight, pointed, White, 115 mm
Catalog # 231-0247
Supplier:   Brand
Forceps
Catalog # 231-0247
Supplier:  Brand
Forceps
Forceps

Tweezers, in PMP, Straight, pointed, White, 115 mm

Price
23,10 €
Log in for availability
1 * 10 unid.
Product Details & Documents

PMP, white.

  • Pointed ends
  • Autoclavable (121 °C)
Specifications
  • Type:
    Straight, pointed
  • Colour:
    White
  • Description:
    Tweezers, in PMP
  • Length:
    115 mm