Laboratory bottles, narrow neck, with thread
250 ml, GL 32, 160 mm, 65×65×160 mm, Amber
Catalog # BRAA704014
Supplier: Brand
Jump to:
Product Details & Documents
Soda-lime glass, amber, coated or uncoated.
- Good chemical and physical properties
- Maximum operating temperature for coated bottles is 80 °C
- To preserve the coating, do not clean at temperatures exceeding 60 °C
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials. The coating envelops the glass bottle like a protection skin. Should the bottle break, the splintering hazards are reduced considerably.
Documents
Brand Dispensette® S
Product Literature
BRND704010_TDS_EN BRND704020_TDS_EN
Datasheet
GA Dispensette S
Manual
Specifications
- Thread:GL 32
- Capacity:250 ml
- Colour:Amber
- Height:160 mm
- Seq:07
- W×D×H:65×65×160 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



