Container, straight, capacity 40ml, naterial PS natural, height 70mm, diameter 30mm, screw cap white color. 1 * 1.000 items
Catalog # 216-2685
Supplier: VWR Collection
Related Information
216-2693
Images 360
TDS_VWR_Straight_Sample_Containers_PS_with_Screw_Cap_Aseptic.pdf
Product specification
Frequently Bought Together