Order Entry
Portugal
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Gouge, inclined forward, Wagner, 15 mm, 340 mm
Catalog # AESCND122R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCND122R
Supplier:  Aesculap
Gouges, Aesculap®

Gouge, inclined forward, Wagner, 15 mm, 340 mm

Price
Inquire For Price
Log in for availability
1 * 1 unid.
Product Details & Documents
Specifications
  • Type:
    Wagner
  • Width:
    15 mm
  • Description:
    Gouge, inclined forward
  • Length:
    340 mm
  • Seq:
    46
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.