Order Entry
Portugal
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Lamina gouge, cranked, Capener, 8 mm, 235 mm
Catalog # AESCFL438R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL438R
Supplier:  Aesculap
Gouges, Aesculap®

Lamina gouge, cranked, Capener, 8 mm, 235 mm

Price
Inquire For Price
Log in for availability
1 * 1 unid.
Product Details & Documents
Specifications
  • Type:
    Capener
  • Width:
    8 mm
  • Description:
    Lamina gouge, cranked
  • Length:
    235 mm
  • Seq:
    51
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.