Order Entry
Portugal
Orders LinkContactUsLinkComponent
Gouges, Aesculap®

Gouges, Aesculap®

Spongiosa gouge, Lexer, 10 mm, 280 mm
Catalog # AESCFL423R
Supplier:   Aesculap
Gouges, Aesculap®
Catalog # AESCFL423R
Supplier:  Aesculap
Gouges, Aesculap®

Spongiosa gouge, Lexer, 10 mm, 280 mm

Price
Inquire For Price
Log in for availability
1 * 1 unid.
Product Details & Documents
Specifications
  • Type:
    Lexer
  • Width:
    10 mm
  • Description:
    Spongiosa gouge
  • Length:
    280 mm
  • Seq:
    57
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.