Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dissecting scissors, Aesculap® Durotip®
Dissecting scissors, Aesculap® Durotip®
Curved, blunt, wave cut, 145 mm
Catalog # AESCBC259W
Supplier:   Aesculap
Dissecting scissors, Aesculap® Durotip®
Catalog # AESCBC259W
Supplier:  Aesculap
Dissecting scissors, Aesculap® Durotip®
Curved, blunt, wave cut, 145 mm
Price
241,00 €
Log in for availability
1 * 1 unid.
Product Details & Documents
Dissecting scissor for dental applications. Type Baby-Metzenbaum.
  • With inserted carbide cutting edges
  • Excellent cutting accuracy
  • Long working life
  • Gold-plated rings
Wave cut models prevent tissue from slipping out during the cutting process.
Specifications
  • Type:
    Curved, blunt, wave cut
  • Length:
    145 mm
  • Seq:
    04