Blanking flanges
Blanking flanges NW, DN 16, Aluminium
Catalog # EDWAC10512368
Supplier: EDWARDS HIGH VACUUM METTLER
Jump to:
Product Details & Documents
Blanking flanges made of aluminium or stainless steel.
Used for vacuum system.
Specifications
- Description:Blanking flanges NW, DN 16
- Material:Aluminium
- Seq:01