Clamps, micro
Clamps, micro, finger with cork, round jaws, Stainless steel 18/10, 15 mm, 150 mm
Catalog # BOCH5400
Supplier: BOCHEM
Jump to:
Product Details & Documents
Micro clamps made of stainless steel.
- Fingers are covered with either a layer of cork or with silicone coating to protect clamped items
- With round or angled jaws
Specifications
- Spanning width:15 mm
- Description:Clamps, micro, finger with cork, round jaws
- Length:150 mm
- Material:Stainless steel 18/10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



