Rubber washers
Washer, I.Ø×H 35×7 mm, 75 mm
Catalog # DENE2403575
Supplier: DEUTSCH NEUMANN
Jump to:
Product Details & Documents
These washers are made from red natural rubber.
- Hardness 40 Shore A
Specifications
- Ø:75 mm
- Description:Washer, I.Ø×H 35×7 mm
- Seq:03
Recomendações
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



