Rubber washers
Washer, I.Ø×H 12×5 mm, 65 mm
Catalog # DENE2401265
Supplier: DEUTSCH NEUMANN
Jump to:
Product Details & Documents
These washers are made from red natural rubber.
- Hardness 40 Shore A
Specifications
- Ø:65 mm
- Description:Washer, I.Ø×H 12×5 mm
- Seq:01



