Entrada de Pedido
Portugal
Orders LinkContactUsLinkComponent
Snap-off replacement blades for cutters
Snap-off replacement blades for cutters
Spare cutter blades, 10 snap-off blades, 9 mm, in plastic box, Aço de carbono
Catalog # 231-2902
Supplier:   WEDO WERNER DORSCH
Snap-off replacement blades for cutters
Catalog # 231-2902
Supplier:  WEDO WERNER DORSCH
Snap-off replacement blades for cutters
Snap-off replacement blades for cutters
Spare cutter blades, 10 snap-off blades, 9 mm, in plastic box, Aço de carbono
Price
4,60 €
Log in for availability
1 * 10 unid.
Product Details & Documents
These snap-off replacement cutter blades are made of SK5 carbon steel and are suitable for almost all commercial cutter models.
  • High quality
  • High edge retention and sharpness
Packaging: Supplied in a plastic box with 10 blades.
Specifications
  • Descrição:
    Spare cutter blades, 10 snap-off blades, 9 mm, in plastic box
  • Material:
    Aço de carbono
  • Seq:
    02