Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Substrate buffers, Pierce™

Pierce™ Diethanolamine Substrate buffer, 5X, 225 ml
Catalog # PIER34064
Substrate buffers, Pierce™
Catalog # PIER34064
CAS Number:  
Substrate buffers, Pierce™
Substrate buffers, Pierce™

Pierce™ Diethanolamine Substrate buffer, 5X, 225 ml

CAS Number:  
Price
108,00 €
Log in for availability
1 * 225 mL
Product Details & Documents

These stable peroxide and diethanolamine substrate buffers produce optimised, reliable and convenient stock solutions for HRP and alkaline phosphatase assays detection respectively.

  • Ready to use
  • Minimises assay-to-assay variability
Specifications

Specifications

  • Pk:
    225 ml
  • Description:
    Pierce™ Diethanolamine Substrate buffer, 5X

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.