Order Entry
Portugal
Orders LinkContactUsLinkComponent
Hydrogen peroxide 35%, J.T.Baker®

Hydrogen peroxide 35%, J.T.Baker®

Plastic bottle, 1 L
Catalog # 7048.1000
Oxidizing
Irritant
Corrosive
Toxic
Health
Supplier:   Avantor
Hydrogen peroxide 35%, J.T.Baker®
Catalog # 7048.1000
Supplier:  Avantor
CAS Number:  7722-84-1
Hydrogen peroxide 35%, J.T.Baker®

Plastic bottle, 1 L

CAS Number:  7722-84-1
Price
35,06 €
Log in for availability
1 * 1 L
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7722-84-1
  • Formula:H₂O₂
  • MDL Number:MFCD00011333
  • Molecular Weight:34.01 g/mol
  • Boiling Pt:∼107 °C (1013 hPa)
  • Melting Pt:< 0 °C
  • Density:1.11 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-765-0
  • UN:2014
  • ADR:5.1,II
  •  :Density: 1.11 g/cm³ (20 °C)
Specifications

Specification Test Results

  • Assay *:
    min. 35 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.