Order Entry
Portugal
Orders LinkContactUsLinkComponent
Silver nitrate standardised solution 1.0 N in aqueous solution

Silver nitrate standardised solution 1.0 N in aqueous solution

Glass bottle, 500 mL
Catálogo # 35597.AP
Corrosivo
Ambiente
Silver nitrate standardised solution 1.0 N in aqueous solution
Catálogo # 35597.AP
Número CAS:  7761-88-8
Silver nitrate standardised solution 1.0 N in aqueous solution
Silver nitrate standardised solution 1.0 N in aqueous solution

Glass bottle, 500 mL

Número CAS:  7761-88-8
Price
812,00 €
Iniciar Sessão para disponibilidade
1 * 500 mL
Product Details & Documents

CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless UN No.: 3264 MDL No.: MFCD00003414

Sinônimos:  Lunar caustic, Silver mononitrate, Nitric acid silver salt
  • Número CAS:7761-88-8
  • Fórmula:AgNO₃
  • Número MDL:MFCD00003414
  • Molecular Weight:169.87 g/mol
  • Temperatura de Armazenamento:Ambient
  • EINECS:231-853-9
  • UN:3264
  • ADR:8,II
Especificações

Especificações

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Especificações

Recomendações

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.