Order Entry
Portugal
Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)

Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 400 mL
Catalog # PROMV4233
Supplier:   Promega
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
Catalog # PROMV4233
Supplier:  Promega
CAS Number:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)

EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 400 mL

CAS Number:  
Price
151,00 €
Log in for availability
1 * 400 mL
Product Details & Documents

This EDTA solution is a chelator of divalent cations and is suitable for biochemistry and molecular biology applications

  • Available in 0,5 M or 10 mM concentrations
  • 10 mM concentration also provided as part of Maxwell® HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)

It is supplied as a solution in deionized water.

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    400 mL
  • Description:
    EDTA buffer solution pH 8,0 (0,5 mol/l)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.