Urea
Glass bottle for solids, 1 kg
Catalog # PROMV31711000
Supplier: Promega
Jump to:
Product Details & Documents
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
Specifications
Specification Test Results
- Purity:≥99.0%
- Melting Point:132–135°C
- A280 at 8M in water:≤0.10
- Chloride:≤0.0005%
- Heavy Metals:≤0.001%
- Iron:≤0.001%
- Cyanate:none detected
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg



