Order Entry
Portugal
Orders LinkContactUsLinkComponent
T7 RNA polymerase

T7 RNA polymerase

T7 RNA Polymerase (10 - 20 U/µl), 5.000 EU
Catalog # PROMP20775000
Supplier:   Promega
T7 RNA polymerase
Catalog # PROMP20775000
Supplier:  Promega
CAS Number:  
T7 RNA polymerase
T7 RNA polymerase

T7 RNA Polymerase (10 - 20 U/µl), 5.000 EU

CAS Number:  
Price
343,00 €
Log in for availability
1 * 5.000 UE
Product Details & Documents

T7 RNA Polymerase is a DNA-dependent RNA polymerase specific for T7 promoter sequences, that exhibits extremely high specificity for its cognate promoter sequences. Only T7 DNA or DNA cloned downstream from a T7 promoter can serve as a template for T7 RNA Polymerase-directed RNA synthesis.

  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (<1% release)
  • Incorporates ³²P, ³³P, ³H and ³⁵S nucleoside triphosphates
  • Source: Recombinant E. coli strain

Applications:
Generation of templates for in vitro translation
Generation of probes for nucleic acid hybridizations
Generation of RNA processing substrates
Generation of antisense RNA

Storage buffer: 20 mM potassium phosphate (pH 7,7 at 25 °C), 1 mM EDTA, 10 mM DTT, 100 mM NaCI, 0,1% (v/v) Triton® X-100 and 50% (v/v) glycerol.

Unit definition: One unit is defined as the amount of enzyme required to catalyze the incorporation of 5 nmol of CTP into acid-insoluble product in 60 minutes at 37 °C in a total volume of 100 μl.

Delivery information: Supplied with DTT (100 mM) and transcription optimized 5X buffer: 200 mM Tris-HCl (pH 7,9 at 25 °C), 30 mM MgCl₂, 10 mM spermidine, 50 mM NaCl.

Specifications

Specifications

  • Pk:
    5.000 EU
  • Description:
    T7 RNA Polymerase (10 - 20 U/µl)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.