Order Entry
Portugal
Orders LinkContactUsLinkComponent
Buffer solutions in sachets, Certipur®, Supelco®

Buffer solutions in sachets, Certipur®, Supelco®

CertiPur® buffer solution, pH 10,00, boric acid, sodium hydroxide, potassium chloride, 10,00, Borate, Sachet, 30×30 ml, 30
Catalog # 1.99004.0001
Supplier:   Merck
Buffer solutions in sachets, Certipur®, Supelco®
Catalog # 1.99004.0001
Supplier:  Merck
CAS Number:  
Buffer solutions in sachets, Certipur®, Supelco®
Buffer solutions in sachets, Certipur®, Supelco®

CertiPur® buffer solution, pH 10,00, boric acid, sodium hydroxide, potassium chloride, 10,00, Borate, Sachet, 30×30 ml, 30

CAS Number:  
Price
104,00 €
Log in for availability
1 * 30 unid.
Product Details & Documents

For calibration of pH meters.

  • Practical 30 ml sachets - one use per sachet - no risk of contamination
  • Solutions are always fresh and ready to use
  • Traceable to SRM from NIST and PTB
  • Reference temperature: +25 °C
  • Precision: Deviation is max. ±0,015 pH units

pH value is measured with a combined glass electrode after 5-point calibration according to DIN 19268 with reference buffer solutions according to DIN 19266, IUPAC, NIST, Ph. Eur. and USP.

Specifications

Specifications

  • Pack type:
    Sachet
  • pH buffer type:
    Borate
  • Pk:
    30
  • Description:
    CertiPur® buffer solution, pH 10,00, boric acid, sodium hydroxide, potassium chloride
  • Pk Info:
    30×30 ml
  • pH value:
    10,00
  • Cat. No.:
    1.99004.0001
  • M-Clarity quality level:
    MQ100

Specifications