Ammonium acetate, Supelco®
Plastic bottle, 1 kg
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Merck Index:13,00495
- Melting Pt:114 °C
- Density:1.17 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Flash Pt:136 °C
- Storage Temperature:Ambient
- EINECS:211-162-9
Specifications
Specification Test Results
- Solubility:1480 g/l (4 °C)
- Melting Point:114 °C
- Molar Mass:77.08 g/mol
- Bulk Density:410 kg/m3
- Density:1.17 g/cm3 (20 °C)
- pH:6.7 - 7.3 (50 g/l, H2O, 25 °C)
- Storage:Store at +15°C to +25°C.
- Hill Formula:C2H7NO2
- Chemical Formula:CH3COONH4
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



