Order Entry
Portugal
ContactUsLinkComponent
TitroLine easy module 3. Basic unit without electrode, ready to use with 20 ml dosing unit TZ 3130 with tubes and screw threads, mag netic stirrer TM 96, titration tip with tube, electrode-/ titration tip holder TZ 3660, stand rod TZ 3665, 230 V wit 1 * 1 items
Catalog # 661-0063
Supplier:  SI Analytics
TitroLine easy module 3. Basic unit without electrode, ready to use with 20 ml dosing unit TZ 3130 with tubes and screw threads, mag netic stirrer TM 96, titration tip with tube, electrode-/ titration tip holder TZ 3660, stand rod TZ 3665, 230 V wit 1 * 1 items
Catalog # 661-0063
Supplier:  SI Analytics
Frequently Bought Together