Order Entry
Portugal
Orders LinkContactUsLinkComponent
Büchner funnels

Büchner funnels

235 ml, 80 mm
Catalog # 511-0008
Supplier:   KARTELL
Büchner funnels
Catalog # 511-0008
Supplier:  KARTELL
Büchner funnels
Büchner funnels

235 ml, 80 mm

Price
17,00 €
Log in for availability
1 * 1 unid.
Product Details & Documents

PP, white.

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    235 ml
  • For filter Ø:
    80 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.