Order Entry
Portugal
Orders LinkContactUsLinkComponent
Bulk packaged serological pipettes, Falcon®

Bulk packaged serological pipettes, Falcon®

1 ml, 0,01 ml, +, 25 per bag
Catalog # 734-0335
Supplier:   Corning
Bulk packaged serological pipettes, Falcon®
Catalog # 734-0335
Supplier:  Corning
Bulk packaged serological pipettes, Falcon®
Bulk packaged serological pipettes, Falcon®

1 ml, 0,01 ml, +, 25 per bag

Price
255,52 €
Log in for availability
1 * 1.000 unid.
Product Details & Documents

The pipettes graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.

  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0,1 EU/ml
  • Color-coded package and markings for ease of identification

Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduations should face you every time.

Specifications
  • Packed:
    25 per bag
  • Sterile:
    +
  • Capacity:
    1 ml
  • Seq:
    01
  • Division:
    0,01 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.