Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone 97%

Dapsone 97%

Glass bottle for solids, 100 g
Catalog # ACRO104361000
Irritant
Dapsone 97%
Catalog # ACRO104361000
CAS Number:  80-08-0
Dapsone 97%
Dapsone 97%

Glass bottle for solids, 100 g

CAS Number:  80-08-0
Price
47,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Melting Pt:173…178 °C
  • EINECS:201-248-4

Documents

Specifications

Specification Test Results

  • Appearance:
    White to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    173 to 178 °C
  • Titration with HClO4:
    ≥96.0 %
  • Apparent density:
    0.3 to 0.5 g/cm3
  • Titration:
    ≥97.5 % w/w (as Oxalate) On dry substance
  • Iron (Fe):
    ≤200 ppm
  • Insoluble matter:
    ≤0.3 % (in HCl)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.