Order Entry
Portugal
Orders LinkContactUsLinkComponent
Sample loops for Valco valves

Sample loops for Valco valves

Sample loop, for Valco W and UW type valves, Stainless steel, 50 µl
Catalog # JOURJR-SLV-76-50
Supplier:   VICI JOUR RESEARCH
Sample loops for Valco valves
Catalog # JOURJR-SLV-76-50
Supplier:  VICI JOUR RESEARCH
Sample loops for Valco valves

Sample loop, for Valco W and UW type valves, Stainless steel, 50 µl

Price
62,30 €
Log in for availability
1 * 1 unid.
Product Details & Documents

These VICI Jour sample loops are made of high quality stainless steel 316.

  • Fits all Valco W and UW type valves
  • High pressure resistance, low permeability
  • Tolerance of ±0,05 mm and pressure rating of <350 bar

They are cut using an electrolytic cutting process, cleaned with ultrasound and high pressure steam to remove contaminants, dried, and capped with softener-free caps to avoid recontamination of the tubing.

Delivery information: Supplied with stainless hex-head nuts and stainless ferrules for reliable connections.

Specifications
  • Capacity:
    50 µl
  • Description:
    Sample loop, for Valco W and UW type valves
  • Material:
    Stainless steel
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.