Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dressing forceps, Semken
Dressing forceps, Semken
Semken type, delicate, straight, 120 mm
Catalog # 232-2110
Supplier:   HAMMACHER KARL
Dressing forceps, Semken
Catalog # 232-2110
Supplier:  HAMMACHER KARL
Dressing forceps, Semken
Dressing forceps, Semken
Semken type, delicate, straight, 120 mm
Price
40,30 €
Log in for availability
1 * 1 unid.
Product Details & Documents
Delicate dressing forceps made of stainless steel.
Specifications
  • Description:
    Semken type, delicate, straight
  • Length:
    120 mm
  • Seq:
    01