Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Tetradecanoic acid 98%

Bottle
Catalog # APOSOR1015184-25KG
New Product
Supplier:   Apollo Scientific
Tetradecanoic acid 98%
Catalog # APOSOR1015184-25KG
Supplier:  Apollo Scientific
New Product
CAS Number:  544-63-8
Tetradecanoic acid 98%

Bottle

CAS Number:  544-63-8
Price
2 540,00 €
Log in for availability
1 * 25 kg
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Tetradecylic acid, Tetradecanoic acid
  • CAS Number:544-63-8
  • Formula:C₁₄H₂₈O₂
  • MDL Number:MFCD00002744
  • Molecular Weight:228,37 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.