Sunifiram ≥98%
Bottle
Catalog # APOSBUP13511-100MG
New Product
Supplier: Apollo Scientific
Sunifiram ≥98%
Catalog # APOSBUP13511-100MG
Supplier: Apollo Scientific
New Product
CAS Number: 314728-85-3
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: Sunifiram
- CAS Number:314728-85-3
- Formula:C₁₄H₁₈N₂O₂
- Molecular Weight:246,3 g/mol
Specifications
Specifications
- Pack type:Bottle



