Order Entry
Portugal
Orders LinkContactUsLinkComponent
Sunifiram ≥98%

Sunifiram ≥98%

Bottle
Catalog # APOSBUP13511-1G
New Product
Supplier:   Apollo Scientific
Sunifiram ≥98%
Catalog # APOSBUP13511-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  314728-85-3
Sunifiram ≥98%

Bottle

CAS Number:  314728-85-3
Price
804,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Sunifiram
  • CAS Number:314728-85-3
  • Formula:C₁₄H₁₈N₂O₂
  • Molecular Weight:246,3 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.