EN4 ≥98%
Bottle
Catalog # APOSBUP08307-50MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: EN4
- CAS Number:1197824-15-9
- Formula:C₂₅H₂₄N₂O₄
- Molecular Weight:416,47 g/mol
Specifications
Specifications
- Pack type:Bottle



