Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Aklomide ≥98%

Bottle
Catalog # APOSBUP17978-1G
New Product
Supplier:   Apollo Scientific
Aklomide ≥98%
Catalog # APOSBUP17978-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  3011-89-0
Aklomide ≥98%

Bottle

CAS Number:  3011-89-0
Price
105,00 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Aklomide
  • CAS Number:3011-89-0
  • Formula:C₇H₅ClN₂O₃
  • Molecular Weight:200,58 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications