Order Entry
Portugal
Orders LinkContactUsLinkComponent
Aklomide ≥98%

Aklomide ≥98%

Bottle
Catalog # APOSBUP17978-500MG
New Product
Supplier:   Apollo Scientific
Aklomide ≥98%
Catalog # APOSBUP17978-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  3011-89-0
Aklomide ≥98%

Bottle

CAS Number:  3011-89-0
Price
72,40 €
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/life science applications.

Synonyms:  Aklomide
  • CAS Number:3011-89-0
  • Formula:C₇H₅ClN₂O₃
  • Molecular Weight:200,58 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications