Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Dapsone

Bottle
Catalog # APOSOR350018-2.5KG
New Product
Supplier:   Apollo Scientific
Dapsone
Catalog # APOSOR350018-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  80-08-0
Dapsone

Bottle

CAS Number:  80-08-0
Price
254,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.