Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Dapsone

Bottle
Catalog # APOSOR350018-100G
New Product
Supplier:   Apollo Scientific
Dapsone
Catalog # APOSOR350018-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  80-08-0
Dapsone

Bottle

CAS Number:  80-08-0
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications