Order Entry
Portugal
Orders LinkContactUsLinkComponent
4-(Trans-4'-N-Pentylcyclohexyl)Cyclohexanone 99%

4-(Trans-4'-N-Pentylcyclohexyl)Cyclohexanone 99%

New Product
Supplier:   Apollo Scientific
4-(Trans-4'-N-Pentylcyclohexyl)Cyclohexanone 99%
Supplier:  Apollo Scientific
New Product
Certificates
4-(Trans-4'-N-Pentylcyclohexyl)Cyclohexanone 99%

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:84868-02-0
  • Formula:C₁₇H₃₀O
  • Molecular Weight:250,42 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 21,80 €
    21,80 €
    1 * 5 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 36,20 €
    36,20 €
    1 * 25 g
       36,20 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 108,00 €
    108,00 €
    1 * 100 g
       108,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 510,00 €
    510,00 €
    1 * 500 g
       510,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 2 000,00 €
    2 000,00 €
    1 * 2,5 kg
       2 000,00 €

    Specifications:
    New Product
    New Product