Sunifiram ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: Sunifiram
- CAS Number:314728-85-3
- Formula:C₁₄H₁₈N₂O₂
- Molecular Weight:246,3 g/mol
4 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: Sunifiram
- CAS Number:314728-85-3
- Formula:C₁₄H₁₈N₂O₂
- Molecular Weight:246,3 g/mol
Product Family Options
Product Information
- Tipo de embalagemAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



