EN4 ≥98%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
Usage: Biochemical/ life science applications.
Synonyms: EN4
- CAS Number:1197824-15-9
- Formula:C₂₅H₂₄N₂O₄
- Molecular Weight:416,47 g/mol
4 Options Available Below
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms: EN4
- CAS Number:1197824-15-9
- Formula:C₂₅H₂₄N₂O₄
- Molecular Weight:416,47 g/mol
Product Family Options
Product Information
- Pack typeAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



