Order Entry
Portugal
Orders LinkContactUsLinkComponent
Cetyldimethylethylammonium Bromide 99%
Cetyldimethylethylammonium Bromide 99%
Cetyldimethylethylammonium Bromide 99%
Usage: Organic building block/reagent
  • Store at 2 to 8 °C
 :  Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide
  •  :124-03-8
  •  :C₂₀H₄₄BrN
  •  :MFCD00011774
  •  :378,47 g/mol
  •  :Refrigerator
  •  :3077
Product Family Options
  • Pack type
  • Bottle
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €
    New Product
    New Product
  • Bottle
    1 * 100 g
    1 * 100 g - 23,20 €
    23,20 €
    1 * 100 g
       23,20 €
    New Product
    New Product
  • Bottle
    1 * 500 g
    1 * 500 g - 113,00 €
    113,00 €
    1 * 500 g
       113,00 €
  • Bottle
    1 * 2,5 kg
    1 * 2,5 kg - 414,00 €
    414,00 €
    1 * 2,5 kg
       414,00 €
    New Product
    New Product
  • Bottle
    1 * 10 kg
    1 * 10 kg - 1 620,00 €
    1 620,00 €
    1 * 10 kg
       1 620,00 €