Order Entry
Portugal
Orders LinkContactUsLinkComponent
Streptozocin ≥95% Pancreatic beta-cell toxin

Streptozocin ≥95% Pancreatic beta-cell toxin

Marketsource
Supplier:   Abcam
Streptozocin ≥95% Pancreatic beta-cell toxin
Supplier:  Abcam
Marketsource
Certificates
Streptozocin ≥95% Pancreatic beta-cell toxin

2 Options Available Below

Product Details & Documents

Selective pancreatic β-cell toxin. DNA alkylating agent. Diabetagen and anticancer activity in vivo.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  1-Methyl-1-nitroso-3-((2S,3R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)urea, Streptozotocin
  • CAS Number:18883-66-4
  • Molecular Weight:265,22 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% Streptozocin
Product Family Options

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 108,00 €
    108,00 €
    1 * 100 mg
       108,00 €

    Specifications:
    Marketsource
    Marketsource

unology General

Product Information

  • Availability
    Price
  • Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - 267,00 €
    267,00 €
    1 * 500 mg
       267,00 €

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.