G-Biosciences TAE Running Buffer (50X)
About this item
The primary use of TAE Running Buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
50X Concentrated solution: 2 M Tris-Acetate, 0,05 M EDTA (pH 8,3)
TAE has a lower buffer capacity than TBE Running Buffer and can easily become exhausted, however linear, double stranded DNA runs faster on TAE.
2 Options Available Below
Product Details & Documents
The primary use of TAE Running Buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
50X Concentrated solution: 2 M Tris-Acetate, 0,05 M EDTA (pH 8,3)
TAE has a lower buffer capacity than TBE Running Buffer and can easily become exhausted, however linear, double stranded DNA runs faster on TAE.
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- R023TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)Plastic bottle1 LLog in for availability1 * 1 L96,10 €
- R024TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)TAE buffer solution 50X concentrate (TRIS-acetate-EDTA buffer)4 LLog in for availability1 * 4 L246,00 €