Order Entry
Portugal
ContactUsLinkComponent
 
Portable Photometer
Catalog # GRANDEN-600
Supplier:  GRANT INSTRUMENTS
Portable Photometer
Catalog # GRANDEN-600
Supplier:  GRANT INSTRUMENTS

Specifications

  • Absorbance range:
    0 - 3,0 Abs (0,001), 0 - 16,00 McF (0 - 16,00) [Absorbance (Abs), McFarland (McF) and resolution]
  • Accuracy:
    ±0,006 @ 1 Abs, ±0,1 @ 0 - 8 McF
  • Light source:
    LED and silicone photodiode (light and detector)
  • Measurement range:
    600 ±10 nm
  • Description:
    Densitometer
  • Electrical:
    External: Input AC 100 - 240 V, 50/60 Hz; output DC 12 V; Input current/power consumption: 12 V, 2,5 W, 0,2 A
  • W×D×H:
    120×145×65 mm
  • Weight:
    0,5 kg

Specifications

About this item

The DEN-600 is a compact, portable, rechargeable battery powered photometer for OD600, McF turbidity and Bradford protein assay methods.

  • An inexpensive alternative to a spectrophotometer, which is commonly used for these applications

  • Battery powered and compact, it can be comfortably located in a biosafety cabinet, anaerobic chamber or quickly relocated
  • The adjustable cell holder accommodates standard 10 mm path cuvettes, round bottom, conical vials or falcon tubes
  • USB connectivity and software allows data transfer, data processing and calculation

Ideal for measuring estimated total number of cells (OD600), turbidity measurement method (McFarland (McF) and protein concentration measurement (Bradford protein assay method).

Common applications include cell concentration measurement, cell growth data estimation, Log phase estimation for microbial cells induction, competent cell preparation, Bradford protein assay method, antibiotic susceptibility testing, and Inhibitory tests.

Vessel type: Cuvettes, round bottom tubes, falcon tubes
PC requirements: Intel/AMD Processor, 1 GB RAM, Windows Vista/7/8/10/11, USB

Ordering information:

McFarland standards in 12 mm and 16 mm also available (must order separately) as well as 16 mm tubes (box of 100).

Delivery:

Supplied with multi plug adapters and DEN software.