Order Entry
Portugal
Orders LinkContactUsLinkComponent
Human Recombinant BOLA1 (from E. coli)

Human Recombinant BOLA1 (from E. coli)

Catalog # ABCAAB139619-100
Marketsource
Supplier:   Abcam
Human Recombinant BOLA1 (from E. coli)
Catalog # ABCAAB139619-100
Supplier:  Abcam
Marketsource
CAS Number:  
Human Recombinant BOLA1 (from E. coli)
Human Recombinant BOLA1 (from E. coli)
CAS Number:  
Price
549,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Recombinant Human BOLA1 protein is a Human Fragment protein, in the 21 to 137 aa range, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, MS.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Form:
    Liquid
  • Protein/peptide name:
    BOLA1
  • Protein synonyms:
    CGI-143|BOLA1|BolA-like protein 1|hBolA
  • Purity:
    >95%
  • Concentration:
    1 mg/ml
  • Source:
    E. coli
  • Biological activity:
    Standard
  • Shipping temperature:
    20
  • Cat. No.:
    ABCAAB139619-100
  • Protein/peptide type:
    Recombinant
  • Storage conditions:
    Store at −20 °C
  • Formulation:
    pH: 8, Constituents: 20% Glycerol (glycerin, glycerine), 1,17% Sodium chloride, 0,32% Tris HCl, 0,02% (R*,R*)-1,4-Dimercaptobutan-2,3-diol
  • Conjugation:
    Unconjugated
  • Tested applications:
    MS, SDS-PAGE
  • Species:
    Human
  • Animal-free:
    N

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.