Order Entry
Portugal
ContactUsLinkComponent
 
 
DL-Carnitine hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
DL-Carnitine hydrochloride ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

 

Frequently Bought Together