Order Entry
Portugal
ContactUsLinkComponent
 
 
(±)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
(±)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

Frequently Bought Together